3,4,5-Trifluoriodbenzol

Lieferant: FLUOROCHEM

Synonyme: 1,2,3-Trifluoro-5-iodobenzene

021385-5G 021385-1G
FLUO021385-5GEA 146 EUR
FLUO021385-5G FLUO021385-1G
3,4,5-Trifluoriodbenzol
3,4,5-Trifluoriodbenzol
Formel: C₆H₂F₃I
Molecular Weight: 257.9800 g/mol
MDL: MFCD03265259
CAS-Nummer: 170112-66-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR