2-Cyan-4-fluorphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 2-Cyano-4-fluorophenylboronic Acid Pinacol Ester, 2-Cyano-4-fluorobenzeneboronic acid pinacol ester

PN-8788-1G PN-8788-5G
COBBPN-8788-1GEA 75.9 EUR
COBBPN-8788-1G COBBPN-8788-5G
2-Cyan-4-fluorphenylboronsäurepinakolester
2-Cyan-4-fluorphenylboronsäurepinakolester
Formel: C₁₃H₁₅BFNO₂
Molecular Weight: 247,07 g/mol
MDL: MFCD09260444
CAS-Nummer: 461451-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR