(4,5-Difluor-2-nitrophenyl)essigsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-(4,5-Difluoro-2-nitrophenyl)acetic acid
AN-1287-5G
AN-1287-1G
COBBAN-1287-5GEA
2030
EUR
COBBAN-1287-5G
COBBAN-1287-1G
(4,5-Difluor-2-nitrophenyl)essigsäure
(4,5-Difluor-2-nitrophenyl)essigsäure
Formel:
C₈H₅F₂NO₄ Molecular Weight: 217,1300 g/mol |
MDL:
MFCD09878318 CAS-Nummer: 1000339-22-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...