2,2-Difluor-2-(4-nitrophenyl)essigsäure

Lieferant: COMBI-BLOCKS

CA-4656-5G CA-4656-1G
COBBCA-4656-5GEA 0 EUR
COBBCA-4656-5G COBBCA-4656-1G
2,2-Difluor-2-(4-nitrophenyl)essigsäure
2,2-Difluor-2-(4-nitrophenyl)essigsäure
Formel: C₈H₅F₂NO₄
MDL: MFCD11504878
CAS-Nummer: 206360-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR