3-Methoxy-4-pyridinboronsäurepinakolester, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
717665-1G
717665-5G
SIAL717665-1GEA
158
EUR
SIAL717665-1G
SIAL717665-5G
3-Methoxy-4-pyridinboronsäurepinakolester, Sigma-Aldrich®
3-Methoxy-4-pyridinboronsäurepinakolester
Formel:
C₁₂H₁₈BNO₃ Molecular Weight: 235,872 g/mol |
MDL:
MFCD13182222 CAS-Nummer: 1243312-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...