5-Methoxy-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-5109-5G PN-5109-1G
COBBPN-5109-5GEA 0 EUR
COBBPN-5109-5G COBBPN-5109-1G
5-Methoxy-3-pyridinboronsäurepinakolester
5-Methoxy-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₃
MDL: MFCD03412068
CAS-Nummer: 445264-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR