5-Methoxy-3-pyridinboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Methoxy-3-pyridylboronic acid Pinacol ester, 3-Methoxypyridine-5-boronic acid Pinacol ester, 3-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(5-Methoxy-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

M-Clarity™ Qualitätsstufe = MQ100

676624-1G 676624-5G
SIAL676624-1GEA 66.1 EUR
SIAL676624-1G SIAL676624-5G
5-Methoxy-3-pyridinboronsäurepinakolester, Sigma-Aldrich®
5-Methoxy-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₈BNO₃
Molecular Weight: 235,0872 g/mol
MDL: MFCD03412068
CAS-Nummer: 445264-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR