5-Fluor-2-methoxy-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 5-Fluoro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

PN-3971-1G PN-3971-250M
COBBPN-3971-1GEA 737 EUR
COBBPN-3971-1G COBBPN-3971-250M
5-Fluor-2-methoxy-3-pyridinboronsäurepinakolester
5-Fluor-2-methoxy-3-pyridinboronsäurepinakolester
Formel: C₁₂H₁₇BFNO₃
Molecular Weight: 253,0800 g/mol
MDL: MFCD08063053
CAS-Nummer: 1083168-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR