4-Butylbenzonitril

Lieferant: Thermo Scientific

TL00747DA TL00747EA TL00747.50
MAYBTL00747DAEA 0 EUR
MAYBTL00747DA MAYBTL00747EA MAYBTL00747.50
4-Butylbenzonitril
4-Butylbenzonitril
Formel: C₁₁H₁₃N
Molecular Weight: 159,23 g/mol
MDL: MFCD00052345
CAS-Nummer: 20651-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR