1,4-Difluor-2-methyl-5-nitrobenzol
Lieferant: BIONET RESEARCH
Synonyme:
2,5-Difluoro-4-nitrotoluene
10R-0045.1
10R-0045.5
10R-0045.500
BIOT10R-0045.1EA
0
EUR
BIOT10R-0045.1
BIOT10R-0045.5
BIOT10R-0045.500
1,4-Difluor-2-methyl-5-nitrobenzol
1,4-Difluoro-2-methyl-5-nitrobenzene
Formel:
C₇H₅F₂NO₂ Molecular Weight: 173.1169 g/mol |
MDL:
MFCD06200905 CAS-Nummer: 141412-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...