3,5-Dibromchinolin 96%

Lieferant: Thermo Scientific
Achtung

H54859.MD H54859.03 H54859.06
H54859.MDEA 31.4 EUR
H54859.MD H54859.03 H54859.06
3,5-Dibromchinolin 96%
3,5-Dibromoquinoline

Purity: 96% CAS No.: 101861-59-0 Molecular Formula: C9H5Br2N Formula Weight: 286.95 MDL No.: MFCD12024477

Formel: C₉H₅Br₂N
Molecular Weight: 286,95 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12024477
CAS-Nummer: 101861-59-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR