2-Ethoxy-8-methylchinolin

Lieferant: COMBI-BLOCKS

PY-1373-5G PY-1373-1G
COBBPY-1373-5GEA 149 EUR
COBBPY-1373-5G COBBPY-1373-1G
2-Ethoxy-8-methylchinolin
2-Ethoxy-8-methylquinoline
Formel: C₁₂H₁₃NO
MDL: MFCD16036158
CAS-Nummer: 1221793-63-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR