Methyl-3-brom-4-chlorbenzoat

Lieferant: COMBI-BLOCKS

CA-4472-5G CA-4472-1G
COBBCA-4472-5GEA 0 EUR
COBBCA-4472-5G COBBCA-4472-1G
Methyl-3-brom-4-chlorbenzoat
Methyl-3-brom-4-chlorbenzoat
Formel: C₈H₆BrClO₂
MDL: MFCD09751943
CAS-Nummer: 107947-17-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR