tert-Butyl-4-brom-2-chlorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 4-bromo-2-chlorobenzoate

CA-4287-5G CA-4287-1G
COBBCA-4287-5GEA 497 EUR
COBBCA-4287-5G COBBCA-4287-1G
tert-Butyl-4-brom-2-chlorbenzoat
tert-Butyl-4-brom-2-chlorbenzoat
Formel: C₁₁H₁₂BrClO₂
MDL: MFCD08703428
CAS-Nummer: 929000-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR