tert-Butyl-2-brom-4-fluorbenzoat

Lieferant: COMBI-BLOCKS

CA-4289-5G CA-4289-25G CA-4289-1G
COBBCA-4289-5GEA 223 EUR
COBBCA-4289-5G COBBCA-4289-25G COBBCA-4289-1G
tert-Butyl-2-brom-4-fluorbenzoat
tert-Butyl-2-brom-4-fluorbenzoat
Formel: C₁₁H₁₂BrFO₂
MDL: MFCD09800899
CAS-Nummer: 951884-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR