Methyl-5-brom-2,3-difluorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 5-bromo-2,3-difluorobenzoate

CA-4694-1G CA-4694-250M
COBBCA-4694-1GEA 1090 EUR
COBBCA-4694-1G COBBCA-4694-250M
Methyl-5-brom-2,3-difluorbenzoat
Methyl-5-brom-2,3-difluorbenzoat
Formel: C₈H₅BrF₂O₂
Molecular Weight: 251,0249 g/mol
MDL: MFCD12026006
CAS-Nummer: 1150163-69-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR