tert-Butyl-4-brom-2-fluorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl 4-bromo-2-fluorobenzoate

CA-4286-5G CA-4286-1G
COBBCA-4286-5GEA 182 EUR
COBBCA-4286-5G COBBCA-4286-1G
tert-Butyl-4-brom-2-fluorbenzoat
tert-Butyl-4-brom-2-fluorbenzoat
Formel: C₁₁H₁₂BrFO₂
Molecular Weight: 275,1100 g/mol
MDL: MFCD08703426
CAS-Nummer: 889858-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR