Methyl-2-brom-5-methylbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-bromo-5-methylbenzoate

CA-4971-5G CA-4971-1G
COBBCA-4971-5GEA 460 EUR
COBBCA-4971-5G COBBCA-4971-1G
Methyl-2-brom-5-methylbenzoat
Methyl-2-brom-5-methylbenzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229,0700 g/mol
MDL: MFCD00144764
CAS-Nummer: 90971-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR