Methyl-3-brom-4-methylbenzoat ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 3-bromo-4-methylbenzoate
B24372.06
B24372.14
B24372.22
B24372.06EA
27.9
EUR
B24372.06
B24372.14
B24372.22
Methyl-3-brom-4-methylbenzoat ≥99%
Methyl-3-brom-4-methylbenzoat
Purity: 99%
CAS No.: 104901-43-1
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
MDL No.: MFCD00144769
Formel:
C₉H₉BrO₂ Molecular Weight: 229,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00144769 CAS-Nummer: 104901-43-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...