Methyl-4-brom-2-(brommethyl)benzoat

Lieferant: CHEMPUR

Synonyme: Methyl 4-bromo-2-bromomethylbenzoate

03-0009.1 03-0009.5
CHMP03-0009.1EA 0 EUR
CHMP03-0009.1 CHMP03-0009.5
Methyl-4-brom-2-(brommethyl)benzoat
Methyl-4-brom-2-(brommethyl)benzoat
Formel: C₉H₈Br₂O₂
Molecular Weight: 307,97 g/mol
MDL: MFCD04114319
CAS-Nummer: 78471-43-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR