Methyl-4-brom-3-methylbenzoat ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 4-bromo-3-methylbenzoate
A19078.06
A19078.14
A19078.06EA
60.9
EUR
A19078.06
A19078.14
Methyl-4-brom-3-methylbenzoat ≥97%
Methyl-4-brom-3-methylbenzoat
Purity: 97%
CAS No.: 148547-19-7
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
Melting Point: 39-44°
Beilstein Reference: 6593534
Formel:
C₉H₉BrO₂ Molecular Weight: 229.07 g/mol Schmelzpunkt: 39…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00673014 CAS-Nummer: 148547-19-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...