Methyl-4-fluor-2-methylbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 4-fluoro-2-methylbenzoate

CA-4770-5G CA-4770-1G
COBBCA-4770-5GEA 48.6 EUR
COBBCA-4770-5G COBBCA-4770-1G
Methyl-4-fluor-2-methylbenzoat
Methyl-4-fluor-2-methylbenzoat
Formel: C₉H₉FO₂
Molecular Weight: 168.16 g/mol
MDL: MFCD06203790
CAS-Nummer: 174403-69-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR