Methyl-3-bromisonicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-bromoisonicotinate

PY-7374-1G PY-7374-25G PY-7374-5G
COBBPY-7374-1GEA 25.5 EUR
COBBPY-7374-1G COBBPY-7374-25G COBBPY-7374-5G
Methyl-3-bromisonicotinat
Methyl-3-bromisonicotinat
Formel: C₇H₆BrNO₂
Molecular Weight: 216,0300 g/mol
MDL: MFCD03788226
CAS-Nummer: 59786-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR