Methyl-2-amino-5-bromisonicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-amino-5-bromo-4-pyridinecarboxylate

PY-7002-5G PY-7002-1G
COBBPY-7002-5GEA 282 EUR
COBBPY-7002-5G COBBPY-7002-1G
Methyl-2-amino-5-bromisonicotinat
Methyl-2-amino-5-bromisonicotinat
Formel: C₇H₇BrN₂O₂
Molecular Weight: 231,0467 g/mol
MDL: MFCD09027294
CAS-Nummer: 882499-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR