5-Chlor-2-ethoxy-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
5-Chloro-2-ethoxypyridine-3-boronic acid
BB-2884-1G
BB-2884-250M
COBBBB-2884-1GEA
864
EUR
COBBBB-2884-1G
COBBBB-2884-250M
5-Chlor-2-ethoxy-3-pyridinboronsäure
5-Chloro-2-ethoxy-3-pyridineboronic acid
Formel:
C₇H₉BClNO₃ |
MDL:
MFCD13195643 CAS-Nummer: 1217500-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...