5-Chlor-2-methoxy-4-pyridinboronsäure

Lieferant: CHEMPUR

Synonyme: 5-Chloro-2-methoxypyridine-4-boronic acid, (5-Chloro-2-methoxypyridin-4-yl)boronic acid

32-0015.500 32-0015.1
CHMP32-0015.500EA 0 EUR
CHMP32-0015.500 CHMP32-0015.1
5-Chlor-2-methoxy-4-pyridinboronsäure
5-Chloro-2-methoxy-4-pyridineboronic acid
Formel: C₆H₇BClNO₃
Molecular Weight: 187.3900 g/mol
MDL: MFCD06658398
CAS-Nummer: 475275-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR