3-Chlor-2-isobutoxy-5-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(5-Chloro-6-isobutoxypyridin-3-yl)boronic acid
BB-5878-1G
BB-5878-250M
COBBBB-5878-1GEA
692
EUR
COBBBB-5878-1G
COBBBB-5878-250M
3-Chlor-2-isobutoxy-5-pyridinboronsäure
3-Chlor-2-isobutoxy-5-pyridinboronsäure
Formel:
C₉H₁₃BClNO₃ Molecular Weight: 229,4700 g/mol |
MDL:
MFCD12546521 CAS-Nummer: 1217500-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...