Methyl-2-chlorisonicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-chloropyridine-4-carboxylate, Methyl 2-chloroisonicotinate

CA-4094-5G CA-4094-25G CA-4094-100G
COBBCA-4094-5GEA 22 EUR
COBBCA-4094-5G COBBCA-4094-25G COBBCA-4094-100G
Methyl-2-chlorisonicotinat
Methyl-2-chlorisonicotinat
Formel: C₇H₆ClNO₂
Molecular Weight: 171,5810 g/mol
MDL: MFCD01765409
CAS-Nummer: 58481-11-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR