Methyl-2-chlorisonicotinat

Lieferant: CHEMPUR

Synonyme: Methyl 2-chloropyridine-4-carboxylate, Methyl 2-chloroisonicotinate

FL13133.POR 013133-25G 013133-5G
CHMPFL13133.POREA 0 EUR
CHMPFL13133.POR FLUO013133-25G FLUO013133-5G
Methyl-2-chlorisonicotinat
Methyl-2-chlorisonicotinat
Formel: C₇H₆ClNO₂
Molecular Weight: 171.5810 g/mol
MDL: MFCD01765409
CAS-Nummer: 58481-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR