Methyl-2-aminoisonicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 2-aminopyridine-4-carboxylate, Methyl 2-aminoisonicotinate

PY-7854-1G PY-7854-25G PY-7854-5G
COBBPY-7854-1GEA 26.2 EUR
COBBPY-7854-1G COBBPY-7854-25G COBBPY-7854-5G
Methyl-2-aminoisonicotinat
Methyl-2-aminoisonicotinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152,15 g/mol
MDL: MFCD04039316
CAS-Nummer: 6937-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR