Methyl-3-aminoisonicotinat 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl 3-aminoisonicotinate, Methyl 3-aminopyridine-4-carboxylate
H34007.03
H34007.09
H34007.03EA
65.4
EUR
H34007.03
H34007.09
Methyl-3-aminoisonicotinat 97%
Methyl-3-aminoisonicotinat
Formel:
C₇H₈N₂O₂ Molecular Weight: 152,20 g/mol Schmelzpunkt: 84…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02082535 CAS-Nummer: 55279-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...