Ethyl-2-methoxyisonicotinat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 2-methoxyisonicotinate

CA-4327-5G CA-4327-25G CA-4327-1G
COBBCA-4327-5GEA 359 EUR
COBBCA-4327-5G COBBCA-4327-25G COBBCA-4327-1G
Ethyl-2-methoxyisonicotinat
Ethyl-2-methoxyisonicotinat
Formel: C₉H₁₁NO₃
Molecular Weight: 181,1900 g/mol
MDL: MFCD11504865
CAS-Nummer: 105596-61-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR