Methyl-2-methoxyisonicotinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 2-methoxy-4-pyridinecarboxylate, Methyl 2-methoxyisonicotinate
CA-4093-25G
CA-4093-1G
CA-4093-5G
COBBCA-4093-25GEA
925
EUR
COBBCA-4093-25G
COBBCA-4093-1G
COBBCA-4093-5G
Methyl-2-methoxyisonicotinat
Methyl-2-methoxyisonicotinat
Formel:
C₈H₉NO₃ Molecular Weight: 167,1620 g/mol |
MDL:
MFCD08436071 CAS-Nummer: 26156-51-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...