Methyl-2-hydroxyisonicotinat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 1,2-dihydro-2-oxopyridine-4-carboxylate
PY-7078-5G
PY-7078-1G
COBBPY-7078-5GEA
130
EUR
COBBPY-7078-5G
COBBPY-7078-1G
Methyl-2-hydroxyisonicotinat
Methyl-2-hydroxyisonicotinat
Formel:
C₇H₇NO₃ Molecular Weight: 153,1400 g/mol |
MDL:
MFCD09258780 CAS-Nummer: 89937-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...