4-Amino-1-ethyl-1H-pyrazol
Lieferant: SIA ENAMINE
Synonyme:
1-Ethyl-1H-pyrazol-4-amine
EN300-36278.1G
EN300-36278.10
ENAMEN300-36278.1GEA
0
EUR
ENAMEN300-36278.1G
ENAMEN300-36278.10
4-Amino-1-ethyl-1H-pyrazol
4-Amino-1-ethyl-1H-pyrazole
Formel:
C₅H₉N₃ Molecular Weight: 111,1451 g/mol |
MDL:
MFCD05861667 CAS-Nummer: 876343-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...