4-(Methylthio)benzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

279684-25G 279684-5G
SIAL279684-25GEA 206 EUR
SIAL279684-25G SIAL279684-5G
4-(Methylthio)benzonitril, Sigma-Aldrich®
4-(Methylthio)benzonitril
Formel: C₈H₇NS
MDL: MFCD00010381
CAS-Nummer: 21382-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR