3-Chlor-2-fluor-5-(trifluormethyl)phenylacetonitril
Lieferant: FLUOROCHEM
005693-1G
005693-5G
FLUO005693-1GEA
210
EUR
FLUO005693-1G
FLUO005693-5G
3-Chlor-2-fluor-5-(trifluormethyl)phenylacetonitril
3-Chlor-2-fluor-5-(trifluormethyl)phenylacetonitril
Formel:
C₉H₄ClF₄N |
MDL:
MFCD01631515 CAS-Nummer: 261763-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...