2-Brom-N-(2,6-dimethylphenyl)benzamid

Lieferant: COMBI-BLOCKS

CS-9437-1G CS-9437-5G
COBBCS-9437-1GEA 98.3 EUR
COBBCS-9437-1G COBBCS-9437-5G
2-Brom-N-(2,6-dimethylphenyl)benzamid
2-Brom-N-(2,6-dimethylphenyl)benzamid
Formel: C₁₅H₁₄BrNO
MDL: MFCD00027948
CAS-Nummer: 195383-89-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR