2-Brom-N-(3,4-dimethylphenyl)benzamid

Lieferant: COMBI-BLOCKS

CS-9438-5G CS-9438-1G
COBBCS-9438-5GEA 303 EUR
COBBCS-9438-5G COBBCS-9438-1G
2-Brom-N-(3,4-dimethylphenyl)benzamid
2-Brom-N-(3,4-dimethylphenyl)benzamid
Formel: C₁₅H₁₄BrNO
MDL: MFCD00783940
CAS-Nummer: 303991-53-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR