2-Amino-N-(4-methylphenyl)benzamid

Lieferant: COMBI-BLOCKS

Synonyme: 2-Amino-N-(p-tolyl)benzamide

CA-4528-1G CA-4528-5G
COBBCA-4528-1GEA 98.3 EUR
COBBCA-4528-1G COBBCA-4528-5G
2-Amino-N-(4-methylphenyl)benzamid
2-Amino-N-(4-methylphenyl)benzamid
Formel: C₁₄H₁₄N₂O
Molecular Weight: 226,2700 g/mol
MDL: MFCD00115975
CAS-Nummer: 32212-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR