2-(Chlormethyl)-5-nitrobenzoxazol

Lieferant: BIONET RESEARCH

6X-0702.500 6X-0702.1 6X-0702.5 6X-0702.10
BIOT6X-0702.500EA 0 EUR
BIOT6X-0702.500 BIOT6X-0702.1 BIOT6X-0702.5 BIOT6X-0702.10
2-(Chlormethyl)-5-nitrobenzoxazol
2-(Chloromethyl)-5-nitrobenzoxazole
Formel: C₈H₅ClN₂O₃
MDL: MFCD01596292
CAS-Nummer: 119198-10-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR