2-(Chlormethyl)-6-nitrobenzoxazol

Lieferant: BIONET RESEARCH

6X-0703.500 6X-0703.1 6X-0703.5 6X-0703.10
BIOT6X-0703.500EA 0 EUR
BIOT6X-0703.500 BIOT6X-0703.1 BIOT6X-0703.5 BIOT6X-0703.10
2-(Chlormethyl)-6-nitrobenzoxazol
2-(Chloromethyl)-6-nitrobenzoxazole
Formel: C₈H₅ClN₂O₃
MDL: MFCD00775454
CAS-Nummer: 221638-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR