2-Chlor-4-formylpyridin Hydrat

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloroisonicotinaldehyde hydrate(contain dehydrate aldehyde)

PY-1459-1G PY-1459-25G PY-1459-5G
COBBPY-1459-1GEA 25.9 EUR
COBBPY-1459-1G COBBPY-1459-25G COBBPY-1459-5G
2-Chlor-4-formylpyridin Hydrat
2-Chlor-4-formylpyridin Hydrat
Formel: C₆H₄ClNO·H₂O
Molecular Weight: 159,57 g/mol
MDL: MFCD16618978
CAS-Nummer: 1228957-10-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR