1-(2-Aminoethyl)piperidin 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-(2-Aminoethyl)piperidine

A18373.06 A18373.14
A18373.06EA 45.9 EUR
A18373.06 A18373.14
1-(2-Aminoethyl)piperidin 98%
1-(2-Aminoethyl)piperidin

Purity: 98% CAS No.: 27578-60-5 Molecular Formula: C7H16N2 Formula Weight: 128.22 Flash Point: 57°(134°F) Refractive Index: 1.4740 Boiling Point: 185-186° Beilstein Reference: 103469 UN No.: 2733 MDL No.: MFCD00006516

Formel: C₇H₁₆N₂
Molecular Weight: 128,22 g/mol
Siedepunkt: 185…186 °C
Dichte: 0,903
Flammpunkt: 57 °C (134 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006516
CAS-Nummer: 27578-60-5
EINECS: 248-540-8
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR