Methyl-5-amino-2-brom-4-chlorbenzoat

Lieferant: COMBI-BLOCKS

AN-1448-5G AN-1448-1G
COBBAN-1448-5GEA 0 EUR
COBBAN-1448-5G COBBAN-1448-1G
Methyl-5-amino-2-brom-4-chlorbenzoat
Methyl-5-amino-2-brom-4-chlorbenzoat
Formel: C₈H₇BrClNO₂
MDL: MFCD12546470
CAS-Nummer: 929524-50-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR