Methyl-3-amino-4-chlorbenzoat

Lieferant: COMBI-BLOCKS

Synonyme: Methyl 3-amino-4-chlorobenzoate, Methyl 3-Amino-4-chlorobenzoate, 3-amino-4-chlorobenzoic acid methylester

CA-4684-1G CA-4684-25G CA-4684-5G
COBBCA-4684-1GEA 0 EUR
COBBCA-4684-1G COBBCA-4684-25G COBBCA-4684-5G
Methyl-3-amino-4-chlorbenzoat
Methyl-3-amino-4-chlorbenzoat
Formel: C₈H₈ClNO₂
Molecular Weight: 185,6100 g/mol
MDL: MFCD03470533
CAS-Nummer: 40872-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR