Methyl-5-amino-2-chlorbenzoat

Lieferant: CHEMPUR

Synonyme: Methyl 5-amino-2-chlorobenzoate

034845.1 034845-5G
CHMP034845.1EA 0 EUR
CHMP034845.1 FLUO034845-5G
Methyl-5-amino-2-chlorbenzoat
Methyl-5-amino-2-chlorbenzoat
Formel: C₈H₈ClNO₂
Molecular Weight: 185,6100 g/mol
MDL: MFCD07800315
CAS-Nummer: 42122-75-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR