Methyl-4-amino-3-chlorbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 4-amino-3-chlorobenzoate

M-Clarity™ Qualitätsstufe = MQ100

655902-1G 655902-5G
SIAL655902-1GEA 55.1 EUR
SIAL655902-1G SIAL655902-5G
Methyl-4-amino-3-chlorbenzoat, Sigma-Aldrich®
Methyl-4-amino-3-chlorbenzoat
Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
MDL: MFCD01320686
CAS-Nummer: 84228-44-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR