Methyl-2-amino-3-nitrobenzoat
Lieferant: FLUOROCHEM
Synonyme:
Methyl 2-amino-3-nitrobenzoate
008931-1G
008931-5G
FLUO008931-1GEA
31.2
EUR
FLUO008931-1G
FLUO008931-5G
Methyl-2-amino-3-nitrobenzoat
Methyl-2-amino-3-nitrobenzoat
Formel:
C₈H₈N₂O₄ Molecular Weight: 196.1600 g/mol |
MDL:
MFCD02093531 CAS-Nummer: 57113-91-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...