Ethyl-4-amino-3,5-diiodbenzoat

Lieferant: Thermo Scientific

RDP00017.1 RDP00017.10 RDP00017.50
MAYBRDP00017.1EA 0 EUR
MAYBRDP00017.1 MAYBRDP00017.10 MAYBRDP00017.50
Ethyl-4-amino-3,5-diiodbenzoat
Ethyl-4-amino-3,5-diiodbenzoat
Formel: C₉H₉I₂NO₂
Molecular Weight: 416,982 g/mol
MDL: MFCD00017092
CAS-Nummer: 5400-81-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR